Learn More
SMART TUBE INC Standard Smart Tubes MTS1P, case of 100
Catalog No. 501351690
Encompass
Standard Smart Tube, case of one hundred. Optimized to work with 1 milliliter of whole blood. Includes THAWLYSE1 Thaw-Lyse, 1000X Concentrate the buffer used after thawing samples stored frozen in Smart Tubes. Also includes SERT1 a bag of one hundred (100) Processing Inserts used to process the thawed sample. Inquire about Smart Tubes optimized to work with other sample volumes as well as Smart Tubes pre-loaded with biologics or drugs of interest. User information must be added to all orders for accurate processing
This item is not returnable. View return policy
Catalog No. 501351690 Supplier SMART TUBE INC Supplier No. MTS1P
Standard Smart Tube, case of one hundred. Optimized to work with 1 milliliter of whole blood. Includes THAWLYSE1 Thaw-Lyse, 1000X Concentrate the buffer used after thawing samples stored frozen in Smart Tubes. Also includes SERT1 a bag of one hundred (100) Processing Inserts used to process the thawed sample. Inquire about Smart Tubes optimized to work with other sample volumes as well as Smart Tubes pre-loaded with biologics or drugs of interest. User information must be added to all orders for accurate processing