Learn More
Ampco Safety Tools Standard Socket, 1/4 in. Drive (Metric)
SDP
Catalog No. 19174464 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric):
13 mm
16 mm
17 mm
21 mm
22 mm
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-174-464 16 mm
19-174-444 17 mm
19-174-446 17 mm
19-174-447 21 mm
19-174-448 21 mm
19-174-449 22 mm
19-174-452 13 mm
19-174-453 13 mm
19-174-454 13 mm
19-174-456 13 mm
19-174-457 13 mm
19-174-460 13 mm
19-174-461 16 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-174-464 Supplier Ampco Safety Tools Supplier No. SS1/4D9MM
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Standard Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Drive Size 1/4 in.
Length (English) 0.63 in.
Length (Metric) 16 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Product Type Standard Socket Wrench
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.