Learn More
Ampco Safety Tools Standard Socket, 3/8 in. Drive (Metric)
SDP
Catalog No. 19174598 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (Metric):
19 mm
21 mm
22 mm
25 mm
27 mm
30 mm
32 mm
33 mm
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-174-598 21 mm
19-174-580 21 mm
19-174-581 22 mm
19-174-582 25 mm
19-174-583 25 mm
19-174-584 27 mm
19-174-585 27 mm
19-174-586 30 mm
19-174-587 30 mm
19-174-588 30 mm
19-174-589 30 mm
19-174-590 30 mm
19-174-591 32 mm
19-174-592 33 mm
19-174-593 33 mm
19-174-594 33 mm
19-174-595 33 mm
19-174-596 19 mm
19-174-597 19 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-174-598 Supplier Ampco Safety Tools Supplier No. SS3/8D9MM
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Standard Socket performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Drive Size 3/8 in.
Length (English) 0.83 in.
Length (Metric) 21 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Product Type Standard Socket Wrench
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.