Learn More
Myron L Company Conductivity solution 7000 microsiemens
SDP
Catalog No. NC9646369
Encompass_Preferred
Conductivity solution 7000 microsiemens, 5687 PPM gallon size. If a COA is required for this item please also order Fisher part # NC9672510, which is the $57 COA fee. If the COA is requested after the order is placed the fee will increase to $70
Catalog No. NC9646369 Supplier Myron L Company Supplier No. KCL7000G
Conductivity solution 7000 microsiemens, 5687 PPM gallon size. If a COA is required for this item please also order Fisher part # NC9672510, which is the $57 COA fee. If the COA is requested after the order is placed the fee will increase to $70