Learn More
Electron Microscopy Sciences Gilder Standard Square 100 mesh Nickel Grid
SDP
Catalog No. 5029106
Encompass_Preferred
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Nickel
Pitch: 250 µm
Hole: 205 µm
Bar: 45 µm
:
Catalog No. 50-291-06 Supplier Electron Microscopy Sciences Supplier No. G100NI
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Nickel
Pitch: 250 µm
Hole: 205 µm
Bar: 45 µm
:
Product Title