Learn More
Electron Microscopy Sciences Gilder Standard Square 200 mesh Copper Grid
SDP
Catalog No. 5029115
Encompass_Preferred
A combination of thin and thick bar grids, with a mark on the rim for orientation.
Grid: Copper
Pitch: 125 µm
Hole: 95 µm
Bar: 35-25 µm
:
Catalog No. 50-291-15 Supplier Electron Microscopy Sciences Supplier No. G200TTCU
A combination of thin and thick bar grids, with a mark on the rim for orientation.
Grid: Copper
Pitch: 125 µm
Hole: 95 µm
Bar: 35-25 µm
:
Product Title