Learn More
Electron Microscopy Sciences Gilder Standard Square 200 mesh Copper Grid
SDP
Catalog No. 5029109
Encompass_Preferred
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Copper
Pitch: 125 µm
Hole: 90 µm
Bar: 35 µm
:
Catalog No. 50-291-09 Supplier Electron Microscopy Sciences Supplier No. G200CU
Add to Cart
Add to Cart
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Copper
Pitch: 125 µm
Hole: 90 µm
Bar: 35 µm
:
Product Title