Learn More
Electron Microscopy Sciences Gilder Standard Square 200 mesh Nickel Grid
SDP
Catalog No. 5029114
Encompass_Preferred
A thickened version of the standard, G200TH with an assymetric center. A mark on the rim allows for precise orientation of the grids.
Grid: Nickel
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm
:
Catalog No. 50-291-14 Supplier Electron Microscopy Sciences Supplier No. G200THNI
A thickened version of the standard, G200TH with an assymetric center. A mark on the rim allows for precise orientation of the grids.
Grid: Nickel
Pitch: 125 µm
Hole: 85 µm
Bar: 40 µm
:
Product Title