Learn More
Electron Microscopy Sciences Gilder Standard Square 400 mesh Nickel Grid
SDP
Catalog No. 5029095
Encompass_Preferred
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Nickel
Pitch: 62 µm
Hole: 37 µm
Bar: 25 µm
:
Catalog No. 50-290-95 Supplier Electron Microscopy Sciences Supplier No. G400NI
A reliable support specimen grid source. Features well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18 um) thickness.
Grid: Nickel
Pitch: 62 µm
Hole: 37 µm
Bar: 25 µm
:
Product Title