Learn More
Perkin Elmer US LLC Standard Stainless Steel Nebulizer Complete Assembly for PinAAcle
Supplier:  Perkin Elmer US LLC N3160143
Encompass_Preferred
A complete Stainless Steel body with Stainless Steel nebulizer, Stainless Steel needle, and Viton o-rings. It is preferred for general applications with acid concentrations less than 5%, providing exceptional performance and durability. This assembly includes: O-Ring sealing Nebulizer to End Cap, Qty. 1 (09926068) Cap, Qty. 1 (N3161057) Venturi, Qty. 1 (03031810) O-Ring for Venturi, Qty. 1 (09902047) O-Rings for Nebulizer Body, Qty. 2 (09902015) Nebulizer Body, Qty. 1 (N3160133) Nebulizer Insert, Qty. 1 (03031812) O-Ring for Nebulizer Insert, Qty. 1 (09902047) Locking Ring, Qty. 1 (N3161006) Spring, Qty. 1 (03031806) O-Ring for Capillary Needle Assembly, Qty. 1 (09902102) Capillary Needle Assembly with O-Ring, Qty. 1 (03030354) PTFE Washer, Qty. 1 (03031802) Fine Adjustment Nut, Qty. 1 (03031460) Sample Capillary, Qty. 1 (09908265) This nebulizer is compatible with the PerkinElmer PinAAcle 500/900F/900H/900T/900Z AA instruments. Note: This nebulizer ships as standard equipment on ins
Catalog No. 50-211-1348