Learn More
Grainger ASRaymond Gas Spring, Carbon Steel, 60 lbf Force, 30 11/16 in. Extended Length, M8x1.25 Thread Size
Catalog No. 502779014
Encompass_Preferred
This gas spring provides a controlled force of 60 lbf and is designed for reliable performance in industrial applications. Constructed from durable carbon steel, this unit features a 10 in. stroke with an extended length of 30.68 in. (30 11/16 in.) and a compressed length of 20.68 in.
  • 60 lbf force
  • 30.68 in. extended length
  • 20.68 in. compressed length
  • 10 in. stroke
  • M8x1.25 rod thread size
  • 0.866 in. cylinder diameter
  • 0.394 in. rod diameter
  • Carbon steel construction
  • End fittings not included
Catalog No. 50-277-9014 Supplier Grainger Supplier No. 19MU59
This gas spring provides a controlled force of 60 lbf and is designed for reliable performance in industrial applications. Constructed from durable carbon steel, this unit features a 10 in. stroke with an extended length of 30.68 in. (30 11/16 in.) and a compressed length of 20.68 in.
  • 60 lbf force
  • 30.68 in. extended length
  • 20.68 in. compressed length
  • 10 in. stroke
  • M8x1.25 rod thread size
  • 0.866 in. cylinder diameter
  • 0.394 in. rod diameter
  • Carbon steel construction
  • End fittings not included