Learn More
Grainger M-Struts Gas Spring, Carbon Steel, 10 in. Stroke, 278 lbf, 2/Pk.
Catalog No. 502783245
Encompass_Preferred
This standard carbon steel gas spring provides reliable support and motion control for various industrial applications. Designed with fixed resistance, this extension-only damper delivers consistent force and operates effectively in environments up to 400 °F.
  • Constructed from durable carbon steel with a nitride finish
  • Features 10 in. stroke length with 278 lbf force
  • Equipped with M10 x 1.5 threaded rod ends
  • Designed for extension-only damping applications
  • Suitable for high-temperature operation up to 400 °F
  • FDA approved for compliant environments
Catalog No. 50-278-3245 Supplier Grainger Supplier No. 19MU53
This standard carbon steel gas spring provides reliable support and motion control for various industrial applications. Designed with fixed resistance, this extension-only damper delivers consistent force and operates effectively in environments up to 400 °F.
  • Constructed from durable carbon steel with a nitride finish
  • Features 10 in. stroke length with 278 lbf force
  • Equipped with M10 x 1.5 threaded rod ends
  • Designed for extension-only damping applications
  • Suitable for high-temperature operation up to 400 °F
  • FDA approved for compliant environments