Learn More
Grainger Raymond Standard Strut, Carbon Steel, 12 in. Stroke
Catalog No. 502778830
Encompass_Preferred
This standard strut is a carbon steel mechanical compression spring designed for reliable performance in various industrial applications. Featuring a durable nitride finish and a robust construction, this extension-only damper provides controlled support and motion management.
  • Carbon steel construction with nitride finish
  • 12 in. stroke length
  • 63 lbf. force rating
  • Maximum operating temperature of 400 °F
  • 0.500 in. rod diameter
  • 1.25 in. body diameter
  • M8 x 1.25 thread size
Catalog No. 50-277-8830 Supplier Grainger Supplier No. 19MU67
This standard strut is a carbon steel mechanical compression spring designed for reliable performance in various industrial applications. Featuring a durable nitride finish and a robust construction, this extension-only damper provides controlled support and motion management.
  • Carbon steel construction with nitride finish
  • 12 in. stroke length
  • 63 lbf. force rating
  • Maximum operating temperature of 400 °F
  • 0.500 in. rod diameter
  • 1.25 in. body diameter
  • M8 x 1.25 thread size