Learn More
Grainger Raymond Mechanical Compression Spring Strut, Stainless Steel, 12 in Stroke, 21 lbf, 1 ea
Catalog No. 502783246
Encompass_Preferred
These mechanical spring struts are engineered for consistent, repeatable loads and long-term reliability in demanding environments. Utilizing advanced coiled spring technology and steel construction, these units offer an effective alternative to conventional gas springs, providing stable performance across a wide temperature range up to 400°F.
  • Features fixed force resistance of 21 lbf with an initial force of 19 lbf and final force of 26 lbf
  • Constructed with a durable steel cylinder finished in nitride
  • Designed with a 12 in. stroke and 36 19/64 in. extended length
  • Includes M6x1.0 threaded rod
  • Suitable for applications requiring reliable extension-only damping
  • Validated as FDA approved
Catalog No. 50-278-3246 Supplier Grainger Supplier No. 19MU63
These mechanical spring struts are engineered for consistent, repeatable loads and long-term reliability in demanding environments. Utilizing advanced coiled spring technology and steel construction, these units offer an effective alternative to conventional gas springs, providing stable performance across a wide temperature range up to 400°F.
  • Features fixed force resistance of 21 lbf with an initial force of 19 lbf and final force of 26 lbf
  • Constructed with a durable steel cylinder finished in nitride
  • Designed with a 12 in. stroke and 36 19/64 in. extended length
  • Includes M6x1.0 threaded rod
  • Suitable for applications requiring reliable extension-only damping
  • Validated as FDA approved