Learn More
Grainger M-STRUTS Gas Spring, Carbon Steel, 6-19/64 in Extended Length, 6 lbf Force, 1 ea
Catalog No. 502783318
Encompass_Preferred
The standard strut is a precision-engineered carbon steel mechanical counterbalance solution designed for reliable, long-term performance across diverse industries. Built for longevity and consistent operation, it provides repeatable cycles even in demanding environments.
  • Constructed from durable carbon steel with a nitride finish
  • Operates in extreme temperature environments from -40°F to 400°F
  • Features 2 in. stroke and 6 lbf. force
  • Offers high reliability for safety-critical applications
  • Includes M6 x 1 thread size
  • No expiration date
Catalog No. 50-278-3318 Supplier Grainger Supplier No. 19MU07
The standard strut is a precision-engineered carbon steel mechanical counterbalance solution designed for reliable, long-term performance across diverse industries. Built for longevity and consistent operation, it provides repeatable cycles even in demanding environments.
  • Constructed from durable carbon steel with a nitride finish
  • Operates in extreme temperature environments from -40°F to 400°F
  • Features 2 in. stroke and 6 lbf. force
  • Offers high reliability for safety-critical applications
  • Includes M6 x 1 thread size
  • No expiration date