Learn More
Grainger Raymond Mechanical Strut, Carbon Steel, 4 in. Stroke, 45 lbf
Catalog No. 502779428
Encompass_Preferred
These precision-engineered carbon steel mechanical struts provide a highly reliable counterbalance solution designed for long service life, corrosion resistance, and consistent performance across endless cycles and extreme temperature environments. They are suitable for safety-critical applications requiring dependable operation.
  • Carbon steel construction with a nitride finish for durability
  • Fixed force resistance of 45 lbf
  • 4 in. stroke length
  • Operating temperature up to 400 °F
  • M8 x 1.25 rod thread size
  • No expiration date
Catalog No. 50-277-9428 Supplier Grainger Supplier No. 19MU19
These precision-engineered carbon steel mechanical struts provide a highly reliable counterbalance solution designed for long service life, corrosion resistance, and consistent performance across endless cycles and extreme temperature environments. They are suitable for safety-critical applications requiring dependable operation.
  • Carbon steel construction with a nitride finish for durability
  • Fixed force resistance of 45 lbf
  • 4 in. stroke length
  • Operating temperature up to 400 °F
  • M8 x 1.25 rod thread size
  • No expiration date