Learn More
Grainger M-STRUTS Mechanical Gas Spring, Carbon Steel, 4 in. Stroke, 1 ea
Catalog No. 502778826
Encompass_Preferred
This mechanical gas spring utilizes an internal wire compression spring instead of conventional gas and gaskets, allowing for operation in dirty or dusty environments. It provides a regular linear force curve and is ideal for structures subject to high impacts, vibrations, or prolonged static loads. The unit features threads at both ends for flexible installation and compatibility with various end fittings.
  • Designed for high load, impact, and vibration tolerance
  • Operates in a temperature range of -40°C to +200°C
  • Provides a regular linear force curve without final extension dampening
  • Constructed with a black painted steel cylinder
  • Threaded at both ends for flexible mounting
  • Not for use as a mechanical stop
Catalog No. 50-277-8826 Supplier Grainger Supplier No. 19MU23
This mechanical gas spring utilizes an internal wire compression spring instead of conventional gas and gaskets, allowing for operation in dirty or dusty environments. It provides a regular linear force curve and is ideal for structures subject to high impacts, vibrations, or prolonged static loads. The unit features threads at both ends for flexible installation and compatibility with various end fittings.
  • Designed for high load, impact, and vibration tolerance
  • Operates in a temperature range of -40°C to +200°C
  • Provides a regular linear force curve without final extension dampening
  • Constructed with a black painted steel cylinder
  • Threaded at both ends for flexible mounting
  • Not for use as a mechanical stop