Learn More
Grainger M-STRUTS Mechanical Gas Spring, Black Painted Steel, 90 lbf Force, 18-13/16 in. Extended Length, 2/Pk.
Catalog No. 502779316
Encompass_Preferred
This mechanical gas spring utilizes an internal wire compression spring rather than gas or gaskets, allowing for enhanced performance in demanding environments. Constructed from black painted steel, this durable component withstands heavy loads and is resistant to both impact and twisting forces in the longitudinal direction. The unit features M10x1.5 threaded ends for versatile mounting, though end fittings are not included.
  • Features a robust wire spring made of Chrome Silicon Alloy
  • Operates effectively across a temperature range of -40°C to +200°C (-40°F to +400°F)
  • Provides a fixed force of 90 lbf
  • Designed with a 6 in. stroke length and an extended length of 18 13/16 in.
  • Compatible with standard threaded end fittings
  • Suitable for use in applications requiring high load tolerance
Catalog No. 50-277-9316 Supplier Grainger Supplier No. 19MU31
This mechanical gas spring utilizes an internal wire compression spring rather than gas or gaskets, allowing for enhanced performance in demanding environments. Constructed from black painted steel, this durable component withstands heavy loads and is resistant to both impact and twisting forces in the longitudinal direction. The unit features M10x1.5 threaded ends for versatile mounting, though end fittings are not included.
  • Features a robust wire spring made of Chrome Silicon Alloy
  • Operates effectively across a temperature range of -40°C to +200°C (-40°F to +400°F)
  • Provides a fixed force of 90 lbf
  • Designed with a 6 in. stroke length and an extended length of 18 13/16 in.
  • Compatible with standard threaded end fittings
  • Suitable for use in applications requiring high load tolerance