Learn More
Grainger M-STRUTS Mechanical Gas Spring, Black Painted Steel, M10x1.5 Thread, 18 13/16 in Extension, 119 lbf Force, 1 ea
Catalog No. 502782829
Encompass_Preferred
This mechanical gas spring is designed for applications in harsh environments, such as those subject to high impact, vibrations, or dust, where traditional gas-charged springs might fail due to leakage. It utilizes an internal wire compression spring rather than gas or oil to provide a linear force curve. The unit features a robust black painted steel cylinder with a nitride-treated steel piston rod for enhanced corrosion resistance.
  • Features a fixed force of 119 lbf.
  • Equipped with M10x1.5 threaded ends to accommodate various optional end fittings.
  • Supports a wide operating temperature range of -40°C to +200°C (up to 400°F).
  • Provides an extended length of 18 13/16 in. and a stroke length of 6 in.
  • Suitable for long-term load bearing without frequent movement.
Catalog No. 50-278-2829 Supplier Grainger Supplier No. 19MU33
This mechanical gas spring is designed for applications in harsh environments, such as those subject to high impact, vibrations, or dust, where traditional gas-charged springs might fail due to leakage. It utilizes an internal wire compression spring rather than gas or oil to provide a linear force curve. The unit features a robust black painted steel cylinder with a nitride-treated steel piston rod for enhanced corrosion resistance.
  • Features a fixed force of 119 lbf.
  • Equipped with M10x1.5 threaded ends to accommodate various optional end fittings.
  • Supports a wide operating temperature range of -40°C to +200°C (up to 400°F).
  • Provides an extended length of 18 13/16 in. and a stroke length of 6 in.
  • Suitable for long-term load bearing without frequent movement.