Learn More
Grainger M-Struts Gas Spring, Stainless Steel, 6 in. Stroke, 212 lbf, 1 ea.
Catalog No. 502783079
Encompass_Preferred
This standard stainless steel gas spring provides fixed force resistance for controlled movement and support. Designed for durability and consistent performance, it features extension-only damping and is suitable for various applications requiring high-quality mechanical assistance.
  • Constructed from corrosion-resistant stainless steel
  • Fixed force of 212 lbf
  • Stroke length of 6 in.
  • Extended and compressed length of 18 13/16 in.
  • Rod diameter of 0.55 in. and cylinder diameter of 1.1 in.
  • Rod thread size of M10 x 1.5
  • Maximum operating temperature of 400 °F
  • FDA approved
Catalog No. 50-278-3079 Supplier Grainger Supplier No. 19MU35
This standard stainless steel gas spring provides fixed force resistance for controlled movement and support. Designed for durability and consistent performance, it features extension-only damping and is suitable for various applications requiring high-quality mechanical assistance.
  • Constructed from corrosion-resistant stainless steel
  • Fixed force of 212 lbf
  • Stroke length of 6 in.
  • Extended and compressed length of 18 13/16 in.
  • Rod diameter of 0.55 in. and cylinder diameter of 1.1 in.
  • Rod thread size of M10 x 1.5
  • Maximum operating temperature of 400 °F
  • FDA approved