Learn More
Grainger M-Struts Standard Gas Spring, Carbon Steel, 6 in Stroke, 338 lbf
Catalog No. 502779024
Encompass_Preferred
This standard carbon steel gas spring is designed for controlled motion and support applications. It features a fixed resistance and operates as an extension-only damper, providing reliable performance in a variety of mechanical systems.
  • Carbon steel construction for durability
  • Fixed resistance force of 338 lbf
  • Extension-only damping mechanism
  • Stroke length of 6 in.
  • Extended length of 24 13/16 in.
  • Compressed length of 24 51/64 in.
  • M10 x 1.5 rod thread size
  • Maximum operating temperature of 400 °F
  • FDA approved
Catalog No. 50-277-9024 Supplier Grainger Supplier No. 19MU39
This standard carbon steel gas spring is designed for controlled motion and support applications. It features a fixed resistance and operates as an extension-only damper, providing reliable performance in a variety of mechanical systems.
  • Carbon steel construction for durability
  • Fixed resistance force of 338 lbf
  • Extension-only damping mechanism
  • Stroke length of 6 in.
  • Extended length of 24 13/16 in.
  • Compressed length of 24 51/64 in.
  • M10 x 1.5 rod thread size
  • Maximum operating temperature of 400 °F
  • FDA approved