Learn More
Grainger M-STRUTS Gas Spring, Carbon Steel, M6x1.0, 24-11/16 in. Extended Length, 1 ea
Catalog No. 502779258
Encompass_Preferred
This gas spring is a mechanical device that uses pressure from compressed gas and oil to assist in the movement and positioning of covers, windows, and other objects. Constructed from carbon steel with a nitride finish, it provides 27 lbf of fixed resistance force. The cylinder and rod design supports smooth extension in various applications.
  • Extended length of 24 11/16 in
  • Fixed resistance force of 27 lbf
  • Rod thread size of M6x1.0
  • Stroke length of 8 in
  • Maximum operating temperature of 400 °F
  • Cylinder diameter of 0.7 in
  • Rod diameter of 0.31 in
  • Constructed from carbon steel with nitride finish
  • FDA approved
Catalog No. 50-277-9258 Supplier Grainger Supplier No. 19MU45
This gas spring is a mechanical device that uses pressure from compressed gas and oil to assist in the movement and positioning of covers, windows, and other objects. Constructed from carbon steel with a nitride finish, it provides 27 lbf of fixed resistance force. The cylinder and rod design supports smooth extension in various applications.
  • Extended length of 24 11/16 in
  • Fixed resistance force of 27 lbf
  • Rod thread size of M6x1.0
  • Stroke length of 8 in
  • Maximum operating temperature of 400 °F
  • Cylinder diameter of 0.7 in
  • Rod diameter of 0.31 in
  • Constructed from carbon steel with nitride finish
  • FDA approved