Learn More
Grainger M-Struts® Mechanical Gas Spring, Stainless Steel, 10 in. Stroke, 60 lbf Force, 1 ea.
Catalog No. 502782811
Encompass_Preferred
This mechanical spring strut is designed for consistent and repeatable loads in demanding environments. By utilizing a mechanical design rather than pressurized gas or oil, it eliminates the risk of seal failure and leakage common in conventional struts, providing reliable performance in extreme temperature conditions and near caustic substances.
  • Provides 60 lbf of force with fixed resistance
  • Features a 10 in. stroke length
  • Constructed from stainless steel
  • Designed for maintenance-free applications
  • Maximum operating temperature of 400 °F
  • FDA approved
Catalog No. 50-278-2811 Supplier Grainger Supplier No. 19MU60
This mechanical spring strut is designed for consistent and repeatable loads in demanding environments. By utilizing a mechanical design rather than pressurized gas or oil, it eliminates the risk of seal failure and leakage common in conventional struts, providing reliable performance in extreme temperature conditions and near caustic substances.
  • Provides 60 lbf of force with fixed resistance
  • Features a 10 in. stroke length
  • Constructed from stainless steel
  • Designed for maintenance-free applications
  • Maximum operating temperature of 400 °F
  • FDA approved