Learn More
Grainger M-Struts Mechanical Spring Strut, Stainless Steel, 30 13/16 in, 105 lbf, 1 ea
Catalog No. 502783060
Encompass_Preferred
These mechanical spring struts offer a maintenance-free solution for motion control and dampening applications. Designed to function like conventional gas struts without the risks of seal failure or gas and oil leakage, these units provide consistent and repeatable force. They are constructed from stainless steel for enhanced corrosion resistance and are suitable for use in extreme temperature environments, including those up to 400°F.
  • Constructed from durable stainless steel
  • Designed for operation in extreme temperatures up to 400°F
  • Features fixed 105 lbf force and 10 in. stroke length
  • Includes M10x1.5 mounting thread
  • Ideal for use in medical, pharmaceutical, and industrial equipment
Catalog No. 50-278-3060 Supplier Grainger Supplier No. 19MU62
These mechanical spring struts offer a maintenance-free solution for motion control and dampening applications. Designed to function like conventional gas struts without the risks of seal failure or gas and oil leakage, these units provide consistent and repeatable force. They are constructed from stainless steel for enhanced corrosion resistance and are suitable for use in extreme temperature environments, including those up to 400°F.
  • Constructed from durable stainless steel
  • Designed for operation in extreme temperatures up to 400°F
  • Features fixed 105 lbf force and 10 in. stroke length
  • Includes M10x1.5 mounting thread
  • Ideal for use in medical, pharmaceutical, and industrial equipment