Learn More
Grainger M-Struts Gas Spring, Stainless Steel, 63 lbf Force, 12 in. Stroke, 1 ea.
Catalog No. 502783244
Encompass_Preferred
These mechanical struts provide a seal-less, gas-less, and oil-less solution for various motion control and dampening applications. Designed with a robust stainless steel construction, these units deliver consistent loads and long service life even in extreme temperature environments ranging from -40°C to +200°C (up to 400°F).
  • Fixed force of 63 lbf
  • Corrosion-resistant stainless steel construction suitable for FDA applications
  • M8 x 1.25 thread size
  • Stroke length of 12 in.
  • Extended length of 36 11/16 in.
  • Compatible with diverse industries including medical, pharmaceutical, and chemical
  • No hazardous material concerns due to lack of internal seals or gases
Catalog No. 50-278-3244 Supplier Grainger Supplier No. 19MU68
These mechanical struts provide a seal-less, gas-less, and oil-less solution for various motion control and dampening applications. Designed with a robust stainless steel construction, these units deliver consistent loads and long service life even in extreme temperature environments ranging from -40°C to +200°C (up to 400°F).
  • Fixed force of 63 lbf
  • Corrosion-resistant stainless steel construction suitable for FDA applications
  • M8 x 1.25 thread size
  • Stroke length of 12 in.
  • Extended length of 36 11/16 in.
  • Compatible with diverse industries including medical, pharmaceutical, and chemical
  • No hazardous material concerns due to lack of internal seals or gases