Learn More
Grainger M-Struts Standard Gas Spring, Stainless Steel, 21 lbf, 12 in. Stroke, 1 ea.
Catalog No. 502783321
Encompass_Preferred
This standard gas spring utilizes compressed gas and liquid pressure to facilitate the controlled opening and closing of objects such as hatches, covers, and seats. Constructed from durable stainless steel, this mechanical device features a rod and cylinder assembly that provides consistent force during operation. For optimal performance and to ensure the internal components remain properly lubricated, this unit should be installed with the rod in the downward position.
  • Constructed from stainless steel for durability
  • Delivers a fixed force of 21 lbf
  • Features an extended and compressed length of 36 19/64 in.
  • Includes a 12 in. stroke
  • Equipped with an M6x1.0 rod thread size
  • Operates at temperatures up to 400 °F
  • Extension-only damping mechanism
  • End fittings are not included
Catalog No. 50-278-3321 Supplier Grainger Supplier No. 19MU64
This standard gas spring utilizes compressed gas and liquid pressure to facilitate the controlled opening and closing of objects such as hatches, covers, and seats. Constructed from durable stainless steel, this mechanical device features a rod and cylinder assembly that provides consistent force during operation. For optimal performance and to ensure the internal components remain properly lubricated, this unit should be installed with the rod in the downward position.
  • Constructed from stainless steel for durability
  • Delivers a fixed force of 21 lbf
  • Features an extended and compressed length of 36 19/64 in.
  • Includes a 12 in. stroke
  • Equipped with an M6x1.0 rod thread size
  • Operates at temperatures up to 400 °F
  • Extension-only damping mechanism
  • End fittings are not included