Learn More
Grainger M-STRUTS Standard Strut, Stainless Steel, 2 in Stroke, 6 lbf, 1 ea
Catalog No. 502783058
Encompass_Preferred
This standard strut is designed for controlled motion and support, featuring a durable stainless steel construction for reliable performance. It is engineered with a 2 in. stroke and provides fixed resistance, making it suitable for applications requiring precise movement regulation.
  • Constructed from high-quality stainless steel for durability
  • Operates at a maximum temperature of 400°F
  • Features a 0.591 in. cylinder diameter and 0.236 in. rod diameter
  • Includes an M6 x 1 thread size for installation
  • Provides a 2 in. stroke length
Catalog No. 50-278-3058 Supplier Grainger Supplier No. 19MU08
This standard strut is designed for controlled motion and support, featuring a durable stainless steel construction for reliable performance. It is engineered with a 2 in. stroke and provides fixed resistance, making it suitable for applications requiring precise movement regulation.
  • Constructed from high-quality stainless steel for durability
  • Operates at a maximum temperature of 400°F
  • Features a 0.591 in. cylinder diameter and 0.236 in. rod diameter
  • Includes an M6 x 1 thread size for installation
  • Provides a 2 in. stroke length