Learn More
Grainger M-Struts Mechanical Gas Spring, Stainless Steel, 2 in Stroke, 24 lbf
Catalog No. 502779013
Encompass_Preferred
This mechanical gas spring is designed for applications requiring high durability in demanding environments. Unlike conventional gas springs that rely on compressed gas, this device utilizes a wire compression spring, making it suitable for structures subjected to vibrations, impacts, or dirty conditions. It maintains a linear force curve and provides reliable performance even when static for prolonged periods. The cylinder and components are constructed from stainless steel for robust operation.
  • Features a linear force curve for consistent performance
  • Resistant to dust, dirt, and longitudinal impact
  • Ideal for rarely used structures or emergency exits
  • Constructed from stainless steel
  • Operating temperature range of -40°C to +200°C
Catalog No. 50-277-9013 Supplier Grainger Supplier No. 19MU12
This mechanical gas spring is designed for applications requiring high durability in demanding environments. Unlike conventional gas springs that rely on compressed gas, this device utilizes a wire compression spring, making it suitable for structures subjected to vibrations, impacts, or dirty conditions. It maintains a linear force curve and provides reliable performance even when static for prolonged periods. The cylinder and components are constructed from stainless steel for robust operation.
  • Features a linear force curve for consistent performance
  • Resistant to dust, dirt, and longitudinal impact
  • Ideal for rarely used structures or emergency exits
  • Constructed from stainless steel
  • Operating temperature range of -40°C to +200°C