Learn More
Grainger Barnes Group Extension M-Strut, Stainless Steel, 12 11/16 in, 45 lbf, 2/Pk.
Catalog No. 502782796
Encompass_Preferred
This extension gas spring is engineered for superior counterbalancing in applications such as lowering ramps or dropdown hatches. Constructed from durable stainless steel, this maintenance-free component is designed to provide reliable performance in demanding environments with an operating temperature capacity of up to 400 °F.
  • Features a robust stainless steel construction for long-lasting durability
  • Designed for extension-only damping applications
  • Equipped with M8x1.25 rod threading for precise installation
  • Provides a total stroke length of 4 in.
  • Compatible with a maximum operating temperature of 400 °F
  • Supplied without end fittings to allow for application-specific customization
Catalog No. 50-278-2796 Supplier Grainger Supplier No. 19MU20
This extension gas spring is engineered for superior counterbalancing in applications such as lowering ramps or dropdown hatches. Constructed from durable stainless steel, this maintenance-free component is designed to provide reliable performance in demanding environments with an operating temperature capacity of up to 400 °F.
  • Features a robust stainless steel construction for long-lasting durability
  • Designed for extension-only damping applications
  • Equipped with M8x1.25 rod threading for precise installation
  • Provides a total stroke length of 4 in.
  • Compatible with a maximum operating temperature of 400 °F
  • Supplied without end fittings to allow for application-specific customization