Learn More
Grainger M-Struts Gas Spring, Stainless Steel, 12 13/16 in. Extended Length, 75 lbf Force, 1 ea
Catalog No. 502779429
Encompass_Preferred
This gas spring is a mechanical device designed to assist in opening and closing applications such as windows, seats, covers, and hatchbacks by utilizing compressed gas and liquid pressure. The unit features a durable stainless steel construction and provides a fixed force of 75 lbf. Designed for consistent operation, the spring supports extension-only damping and is suitable for environments reaching temperatures up to 400 °F.
  • Fixed force of 75 lbf
  • Constructed from stainless steel
  • Includes M10x1.5 rod threads
  • Extension-only damper type
  • Maximum operating temperature of 400 °F
  • 4 in. stroke length
  • End fittings not included
Catalog No. 50-277-9429 Supplier Grainger Supplier No. 19MU22
This gas spring is a mechanical device designed to assist in opening and closing applications such as windows, seats, covers, and hatchbacks by utilizing compressed gas and liquid pressure. The unit features a durable stainless steel construction and provides a fixed force of 75 lbf. Designed for consistent operation, the spring supports extension-only damping and is suitable for environments reaching temperatures up to 400 °F.
  • Fixed force of 75 lbf
  • Constructed from stainless steel
  • Includes M10x1.5 rod threads
  • Extension-only damper type
  • Maximum operating temperature of 400 °F
  • 4 in. stroke length
  • End fittings not included