Learn More
Grainger M-STRUTS Standard Strut, Stainless Steel, 4 in. Stroke, 21 lbf, 1 ea
Catalog No. 502783221
Encompass_Preferred
This stainless steel gas spring is a standard-type strut designed for applications requiring fixed resistance with a 4 in. stroke. It provides a force of 21 lbf. and is suitable for high-temperature environments.
  • Constructed from durable stainless steel
  • Features a 4 in. stroke length
  • Provides 21 lbf. of force
  • Maximum operating temperature of 400 °F
  • Equipped with M6 x 1.0 rod thread size
  • Designed for extension only
  • FDA approved
Catalog No. 50-278-3221 Supplier Grainger Supplier No. 19MU18
This stainless steel gas spring is a standard-type strut designed for applications requiring fixed resistance with a 4 in. stroke. It provides a force of 21 lbf. and is suitable for high-temperature environments.
  • Constructed from durable stainless steel
  • Features a 4 in. stroke length
  • Provides 21 lbf. of force
  • Maximum operating temperature of 400 °F
  • Equipped with M6 x 1.0 rod thread size
  • Designed for extension only
  • FDA approved