Learn More
Grainger M-STRUTS Gas Spring, Stainless Steel, 119 lbf, 18-13/16 in. Extended Length, 1 ea.
Catalog No. 502783329
Encompass_Preferred
These stainless steel gas springs utilize compressed gas to provide controlled force for assisting in the opening and closing of lids, covers, and hatches. Designed for reliability, these struts deliver fixed resistance and operate effectively at temperatures up to 400 °F.
  • Constructed from durable stainless steel
  • Delivers 119 lbf of force
  • Features 6 in. stroke length
  • Includes M10x1.5 thread size
  • Extension-only damper type
  • Maximum operating temperature of 400 °F
  • Standard design for consistent performance
Catalog No. 50-278-3329 Supplier Grainger Supplier No. 19MU34
These stainless steel gas springs utilize compressed gas to provide controlled force for assisting in the opening and closing of lids, covers, and hatches. Designed for reliability, these struts deliver fixed resistance and operate effectively at temperatures up to 400 °F.
  • Constructed from durable stainless steel
  • Delivers 119 lbf of force
  • Features 6 in. stroke length
  • Includes M10x1.5 thread size
  • Extension-only damper type
  • Maximum operating temperature of 400 °F
  • Standard design for consistent performance