Learn More
Grainger ASRAYMOND Mechanical Strut, Stainless Steel, 6 in Stroke, 15 lbf, 2/Pk.
Catalog No. 502783317
Encompass_Preferred
Mechanical Struts provide a robust, seal-less, gas-less, and oil-less alternative for strut applications. Engineered for performance, these components withstand extreme temperatures and exposure to caustic substances. This unit features fixed resistance and a stainless steel construction, ensuring reliability in demanding environments.
  • Constructed from stainless steel
  • Designed for operation in temperatures up to 400 °F
  • Features 15 lbf force and a 6 in. stroke length
  • Includes M6 x 1.0 thread size for mounting
  • Does not include end fittings
  • Suitable for applications requiring fixed resistance
Catalog No. 50-278-3317 Supplier Grainger Supplier No. 19MU26
Mechanical Struts provide a robust, seal-less, gas-less, and oil-less alternative for strut applications. Engineered for performance, these components withstand extreme temperatures and exposure to caustic substances. This unit features fixed resistance and a stainless steel construction, ensuring reliability in demanding environments.
  • Constructed from stainless steel
  • Designed for operation in temperatures up to 400 °F
  • Features 15 lbf force and a 6 in. stroke length
  • Includes M6 x 1.0 thread size for mounting
  • Does not include end fittings
  • Suitable for applications requiring fixed resistance