Learn More
Grainger M-STRUTS Standard Strut, Stainless Steel, 6 in. Stroke, 338 lbf., 1 ea
Catalog No. 502779025
Encompass_Preferred
This standard strut is engineered for reliable performance in demanding environments, featuring robust stainless steel construction. It provides a fixed force resistance of 338 lbf. and is suitable for high-temperature applications.
  • Constructed from durable stainless steel
  • Fixed force resistance of 338 lbf.
  • Maximum operating temperature of 400°F
  • Stroke length of 6 in.
  • Cylinder diameter of 1.1 in. and rod diameter of 0.55 in.
  • Designed with M10 x 1.5 rod thread size
  • Extension only damper type
  • FDA approved design
Catalog No. 50-277-9025 Supplier Grainger Supplier No. 19MU40
This standard strut is engineered for reliable performance in demanding environments, featuring robust stainless steel construction. It provides a fixed force resistance of 338 lbf. and is suitable for high-temperature applications.
  • Constructed from durable stainless steel
  • Fixed force resistance of 338 lbf.
  • Maximum operating temperature of 400°F
  • Stroke length of 6 in.
  • Cylinder diameter of 1.1 in. and rod diameter of 0.55 in.
  • Designed with M10 x 1.5 rod thread size
  • Extension only damper type
  • FDA approved design