Learn More
Grainger M-STRUTS Gas Spring, Stainless Steel, M6x1.0 Thread, 27 lbf, 18 11/16 in. Extended Length, 2/Pk.
Catalog No. 502782830
Encompass_Preferred
These stainless steel gas springs utilize compressed gas and liquid pressure to assist in the controlled opening and closing of components such as hatches, covers, and seats. The device consists of a rod and piston housed within a cylinder; releasing internal pressure allows the rod to extend and move the attached object. For optimal performance and to prevent drying of the internal liquid, these units must be installed with the rod facing down.
  • Constructed from durable stainless steel
  • Features fixed resistance for consistent operation
  • Provides 27 lbf of force
  • Equipped with M6x1.0 thread size
  • Extended length of 18 11/16 in.
  • Maximum operating temperature of 400 °F
  • Standard extension-only damping
  • FDA approved materials
Catalog No. 50-278-2830 Supplier Grainger Supplier No. 19MU28
These stainless steel gas springs utilize compressed gas and liquid pressure to assist in the controlled opening and closing of components such as hatches, covers, and seats. The device consists of a rod and piston housed within a cylinder; releasing internal pressure allows the rod to extend and move the attached object. For optimal performance and to prevent drying of the internal liquid, these units must be installed with the rod facing down.
  • Constructed from durable stainless steel
  • Features fixed resistance for consistent operation
  • Provides 27 lbf of force
  • Equipped with M6x1.0 thread size
  • Extended length of 18 11/16 in.
  • Maximum operating temperature of 400 °F
  • Standard extension-only damping
  • FDA approved materials