Learn More
Grainger M-STRUTS Gas Spring, Stainless Steel, 57 lbf, 24 11/16 in. Extended Length, 1 ea
Catalog No. 502779311
Encompass_Preferred
This gas spring is a mechanical device engineered to provide controlled assistance in opening and closing heavy objects like lids, hatches, and covers. Utilizing compressed gas within a stainless steel cylinder, the assembly features an 8 in. stroke and delivers a fixed force of 57 lbf. Designed for durability and consistent performance, it is suitable for demanding environments with an operating temperature capacity up to 400 °F.
  • Constructed from stainless steel for enhanced corrosion resistance
  • Features an extended length of 24 11/16 in.
  • Equipped with an M8x1.25 thread size for secure integration
  • Engineered with a 0.86 in. cylinder diameter and 0.39 in. rod diameter
  • Designed for extension-only damper applications
Catalog No. 50-277-9311 Supplier Grainger Supplier No. 19MU48
This gas spring is a mechanical device engineered to provide controlled assistance in opening and closing heavy objects like lids, hatches, and covers. Utilizing compressed gas within a stainless steel cylinder, the assembly features an 8 in. stroke and delivers a fixed force of 57 lbf. Designed for durability and consistent performance, it is suitable for demanding environments with an operating temperature capacity up to 400 °F.
  • Constructed from stainless steel for enhanced corrosion resistance
  • Features an extended length of 24 11/16 in.
  • Equipped with an M8x1.25 thread size for secure integration
  • Engineered with a 0.86 in. cylinder diameter and 0.39 in. rod diameter
  • Designed for extension-only damper applications