Learn More
Grainger M-Struts Gas Spring, Carbon Steel, 24-13/16 in., 96 lbf, 1 ea.
Catalog No. 502779034
Encompass_Preferred
This gas spring is a mechanical device that utilizes compressed gas and liquid pressure to provide assistance in opening and closing objects such as trunks, covers, and hatchbacks. Designed for reliability, this component consists of a piston and rod within a cylinder, facilitating smooth movement as the rod extends when internal pressure is released.
  • Carbon steel construction with a nitride finish
  • Fixed force resistance of 96 lbf
  • 8 in. stroke length
  • M10x1.5 rod thread size
  • Operating temperature up to 400 °F
  • 1.1 in. cylinder diameter and 0.55 in. rod diameter
  • Extended length of 24 13/16 in.
Catalog No. 50-277-9034 Supplier Grainger Supplier No. 19MU52
This gas spring is a mechanical device that utilizes compressed gas and liquid pressure to provide assistance in opening and closing objects such as trunks, covers, and hatchbacks. Designed for reliability, this component consists of a piston and rod within a cylinder, facilitating smooth movement as the rod extends when internal pressure is released.
  • Carbon steel construction with a nitride finish
  • Fixed force resistance of 96 lbf
  • 8 in. stroke length
  • M10x1.5 rod thread size
  • Operating temperature up to 400 °F
  • 1.1 in. cylinder diameter and 0.55 in. rod diameter
  • Extended length of 24 13/16 in.