Learn More
Grainger M-Struts Gas Spring, Stainless Steel, 8 in. Stroke, 18 lbf, 2 ea.
Catalog No. 502783052
Encompass_Preferred
This gas spring is a mechanical device designed to assist in the controlled opening and closing of objects such as hatches, trunks, covers, and seats. Constructed from durable stainless steel, it utilizes internal pressure from compressed gas to provide a fixed resistance of 18 lbf for reliable operation. To ensure longevity and prevent internal fluid drying, it should be installed with the rod oriented downward.
  • Features a 8 in. stroke length and 24-19/64 in. extended length
  • Equipped with a M6x1.0 thread size for secure installation
  • Designed with a 0.59 in. cylinder diameter and 0.23 in. rod diameter
  • Maintains functionality in environments up to a maximum operating temperature of 400 °F
  • Extension-only damper type for smooth, consistent motion
  • FDA approved construction
Catalog No. 50-278-3052 Supplier Grainger Supplier No. 19MU44
This gas spring is a mechanical device designed to assist in the controlled opening and closing of objects such as hatches, trunks, covers, and seats. Constructed from durable stainless steel, it utilizes internal pressure from compressed gas to provide a fixed resistance of 18 lbf for reliable operation. To ensure longevity and prevent internal fluid drying, it should be installed with the rod oriented downward.
  • Features a 8 in. stroke length and 24-19/64 in. extended length
  • Equipped with a M6x1.0 thread size for secure installation
  • Designed with a 0.59 in. cylinder diameter and 0.23 in. rod diameter
  • Maintains functionality in environments up to a maximum operating temperature of 400 °F
  • Extension-only damper type for smooth, consistent motion
  • FDA approved construction