Learn More
Hoefer, Inc Hoefer™ SUB15 Standard Submarine Gel Electrophoresis Unit
SDP
Catalog No. NC2741106
Encompass_Preferred
Provides the most commonly used tray size and comb optionsFeatures and BenefitsThe market standard—a 15 x 15 cm gel tray, with comb options ranging from 1 to 30 samplesMulti-channel pipette compatible combs—with a maximum 30 sample throughput reduce gel loading timeBuffer recirculation ports—may be connected to a peristaltic pump for buffer recirculation during electrophoresis to maintain buffer pH and prevent ionic gradient formationUV-transparent running tray—allows the user to image the gel without risk of damage due to handlingSide handles—for safe and easy transportationIncludes:Buffer Chamber Assembly; Safety Lid; High Voltage Leads; Running Tray with Casting Gates; 16-well combs, 2 pcs; Colored Loading Strips; Buffer Recirculation Ports, 2 pcs;Specifications:Gel Dimensions (W x L): 15 x 15 cmMaximum Buffer Volume: 1.2 LMaximum Power Settings: 300 V, 200 mA, 60 WMaximum Sample Capacity: 120Unit Dimensions (W x L x H): 21.5 x 33.5 x 7 cm
Catalog No. NC2741106 Supplier Hoefer, Inc Supplier No. SUB15
Provides the most commonly used tray size and comb optionsFeatures and BenefitsThe market standard—a 15 x 15 cm gel tray, with comb options ranging from 1 to 30 samplesMulti-channel pipette compatible combs—with a maximum 30 sample throughput reduce gel loading timeBuffer recirculation ports—may be connected to a peristaltic pump for buffer recirculation during electrophoresis to maintain buffer pH and prevent ionic gradient formationUV-transparent running tray—allows the user to image the gel without risk of damage due to handlingSide handles—for safe and easy transportationIncludes:Buffer Chamber Assembly; Safety Lid; High Voltage Leads; Running Tray with Casting Gates; 16-well combs, 2 pcs; Colored Loading Strips; Buffer Recirculation Ports, 2 pcs;Specifications:Gel Dimensions (W x L): 15 x 15 cmMaximum Buffer Volume: 1.2 LMaximum Power Settings: 300 V, 200 mA, 60 WMaximum Sample Capacity: 120Unit Dimensions (W x L x H): 21.5 x 33.5 x 7 cm