Learn More
Bachem H-Leu-Ser-p-nitro-Phe-Nle-Ala-Leu-OMe
SDP
Supplier:  Bachem 4006749.0100
Encompass_Preferred
H-Leu-Ser-p-nitro-Phe-Nle-Ala-Leu-OMe Trifluoroacetate, 99764-63-3, C34H55N7O10, Mr 721.85, 100mg. LSF(NO2)-Nle-AL-Methyl Ester. Standard substrate for chymosin activity determination. This peptide is also a good substrate for pepsin, penicillopepsin, and gastricsin.
Catalog No. 50-194-7027