Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bal Supply Standard Taper Joint Clips

Catalog No. 13684162 Shop All Tip Industries Inc. Products
Change view
Click to view available options
Size:
14
19
24
29
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
13-684-162 24
13-684-160 14
13-684-161 19
13-684-163 29
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-684-162 Supplier Bal Supply Supplier No. 203424
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Bal Supply standard taper joint clips with superior chemical and temperature resistance

  • Nickel Plated Steel
  • Can withstand temperatures up to 932°F
  • For Standard Taper Joints

Specifications

Quantity 10/Pk.
Type Standard Taper Clamp
Material Stainless Steel
For Use With (Equipment) Standard Taper Joints
Size 24
Color Silver
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.