Learn More
Electron Microscopy Sciences Standard Test Specimen Set
SDP
Catalog No. 5029118
Encompass_Preferred
Scanning Electron Microscope with 6 standards (Cobalt and five customer's choices), Faraday cage, BSD Duplex Brass Reference, high or medium resolution grid, specimen for magnification calibration, certificate.
Catalog No. 50-291-18 Supplier Electron Microscopy Sciences Supplier No. 8015400
Scanning Electron Microscope with 6 standards (Cobalt and five customer's choices), Faraday cage, BSD Duplex Brass Reference, high or medium resolution grid, specimen for magnification calibration, certificate.