Learn More
Electron Microscopy Sciences Standard Tip Burnisher
SDP
Catalog No. 5024827
Encompass_Preferred
These burnishers are especially designed for dry transfer products. They are pressure adjustable allowing you to set and maintain the proper pressure for any burnishing style. For maximum adhesion, place the backing sheet over the newly transferred letters and reburnish with the "bone end." The rolling tip burnisher is ideal for large transfers, the standard tip is ideal for standard work, and the fine tip for detail work.
Catalog No. 50-248-27 Supplier Electron Microscopy Sciences Supplier No. 7761501
These burnishers are especially designed for dry transfer products. They are pressure adjustable allowing you to set and maintain the proper pressure for any burnishing style. For maximum adhesion, place the backing sheet over the newly transferred letters and reburnish with the "bone end." The rolling tip burnisher is ideal for large transfers, the standard tip is ideal for standard work, and the fine tip for detail work.