Learn More
Specifications
Specifications
| CAS | 1330-20-7 |
| Quantity | 8 x 10 mL |
| Assay Percent Range | 0.01% |
| Packaging | Glass Ampoule |
| Grade | Karl Fischer |
| Chemical Name or Material | Standard, Water, Apura™ Karl Fischer Water Standard; 0.1mg H2O/g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.