Learn More
Tians Standard-Weight Polypropylene Shoe Covers
Catalog No. 19120871 Shop All Tians International Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-871 Large
19-130-2071 X-Large
19-130-3855 2X-Large
19-120-870 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-120-871 Supplier Tians Supplier No. 514673L
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
35g/m2 fabric is lightweight, durable, and breathable
  • 100% virgin spunbonded, non-linting polypropylene
  • Trimmed in elastic for a snug, adjustable fit
  • Elastic seam is turned in to reduce particle generation
  • With a non-marking, latex-based, anti-skid bottom surface
  • Color: sky blue
Order Info
100 Each per vacuum-packed, heat-sealed polybag; three bags per case.
Compliance
Conform to test standard IES-RP-CC-003.2
Specifications
Size Large
Material Polypropylene
Color Sky Blue
Closure Type Elastic Ankle
Product Type Controlled Environments Shoe Covers