Learn More
Electron Microscopy Sciences MAC X-Ray Fluorescence Single Standard V
SDP
Catalog No. 501900109
Encompass
These standards are supplied as 1/4" (33 mm) diameter pressed pellets or metal foils, and are prepared from selected high purity elements and compounds to ensure interference free-spectra. They are suitable for use by experienced spectroscopists as well as staff training. Each of the pressed pellets is supported by a thin-walled aluminum cup which is protected from damage during handling and minimizes the risk of touching the standard materials (some of which are harmful).
Catalog No. 50-190-0109 Supplier Electron Microscopy Sciences Supplier No. 80093V
These standards are supplied as 1/4" (33 mm) diameter pressed pellets or metal foils, and are prepared from selected high purity elements and compounds to ensure interference free-spectra. They are suitable for use by experienced spectroscopists as well as staff training. Each of the pressed pellets is supported by a thin-walled aluminum cup which is protected from damage during handling and minimizes the risk of touching the standard materials (some of which are harmful).