Learn More
QA Bio Inc QA-Bio, Inc. • Ludger Glucose Homopolymer Ladder 2-AB • 100 picomoles
Supplier:  QA Bio Inc CABGHP30
Encompass
Glucose Homopolymer Ladder • HPLC Calibration Standard • Glucose homopolymer ladder derived from digested wheat starch used as a standard for sizing of glycans • Amount supplied sufficient for 30 HPLC runs.
Catalog No. 50-355-785