Learn More
QA Bio Inc QA-Bio, Inc. • Ludger NA2F Glycan/G2F • 20 µgs
Catalog No. 50355871
Encompass
NA2F Glycan/G2F • Asialo-fucosylated biantennary oligosaccharide • The NA2F glycan (G2F) is an asialo-, bi-antennary complex-type N-linked oligosaccharide. >98% pure as assessed by a combination of 1H-NMR and HPLC. • 20 µgs / 1787 daltons
Catalog No. 50-355-871 Supplier QA Bio Inc Supplier No. CNNA2F20U
NA2F Glycan/G2F • Asialo-fucosylated biantennary oligosaccharide • The NA2F glycan (G2F) is an asialo-, bi-antennary complex-type N-linked oligosaccharide. >98% pure as assessed by a combination of 1H-NMR and HPLC. • 20 µgs / 1787 daltons