Learn More
QA Bio Inc QA-Bio, Inc. • Ludger NGA2 Glycan/G0 • 20 µgs
Catalog No. 50355874
Encompass
NGA2 Glycan/G0 • This NGA2 (G0) glycan is used by biopharmaceutical companies around the world in their analysis of monoclonal antibodies.• >98% pure as assessed by a combination of 1H-NMR and HPLC.
Catalog No. 50-355-874 Supplier QA Bio Inc Supplier No. CNNGA220U
NGA2 Glycan/G0 • This NGA2 (G0) glycan is used by biopharmaceutical companies around the world in their analysis of monoclonal antibodies.• >98% pure as assessed by a combination of 1H-NMR and HPLC.