Learn More
QA Bio Inc QA-Bio, Inc. • Ludger NGA2F Glycan/G0F • 20 µgs
Catalog No. 50355875
Encompass
NGA2F Glycan/G0F • The NGA2F G0F glycan is an asialo-agalacto-fucosylated biantennary oligosaccharide. • >98% pure as assessed by a combination of 1H-NMR and HPLC. • 20 µgs / 1463 daltons
Catalog No. 50-355-875 Supplier QA Bio Inc Supplier No. CNNGA2F20U
NGA2F Glycan/G0F • The NGA2F G0F glycan is an asialo-agalacto-fucosylated biantennary oligosaccharide. • >98% pure as assessed by a combination of 1H-NMR and HPLC. • 20 µgs / 1463 daltons